Description
- A platinum-divinyltetramethylsiloxane complex in a low molecular weight, functional vinyl polymer
Applications
- For RTV and accelerated heat-curing of vinyl-functional silicone systems
- For curing silicone polymer systems to form adhesives, sealant, rubbers and gels due to its reactive nature
- Designed for use with optically clear, high refractive index silicone elastomer systems
Product Details
Property | Average Result |
---|---|
Refractive Index | 1.40 |
% Pt | 2.5 |
Appearance | Light Amber |
Specific Gravity | 0.98 |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.