Description
- A translucent inhibitor known as Ethynlcyclohexanol
- Amounts used can be varied for optimal processing times
Applications
- For use with platinum catalyzed elastomer systems
Product Details
Property | Average Result |
---|---|
Appearance | Translucent Yellow |
Cure | >120 °C |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.