Description
- Peroxide catalyst masterbatch for use with high consistency silicone systems
- Di(2,4-dichlorobenzoyl)peroxide at 50% concentration in silicone oil
Applications
- To vulcanize silicone elastomer systems for calendaring, extrusion, or transfer and compression molding applications
Product Details
Property | Average Result |
---|---|
Appearance | White |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.