Air sampler, MAS-100 NT®
: Merck
The perforated sampling head may be sterilised and can be adjusted to sample horizontally.
- Petri dishes (90 mm) or contact plates (55 mm)
- Programmable delay and individually pre-selected adjustable volumes
- Up to 100 samples can be logged with date, time, user and volume
- Fully automatic calibration cycle, automatic creation of certificate with digital anemometer DA-100 NT® and PC
- Mass flow sensor to ensure a constant airflow rate
- USB port to communicate with a computer
- Autoclavable sampling head to avoid any contamination
Suitable for applications in cleanrooms, pharmaceutical companies, hospitals and the food industry.
: ISO 14698 - 1/2
: Larger sampling heads are available on request.
: Supplied in a case with power supply, USB cable, and CD rom PC software.
- Catalog No:
- 1.17274.0001
- 1.09191.0001
- Gjennomstrømningshastighet:
- 100 L/min
- 100 L/min
- Prøvetakingshode:
- Ø 10 cm including 300 holes of Ø 0,6 mm
- Materiale:
- Aluminium legering
- Aluminium legering
- Alarm:
- Up to 100 events
- Up to 100 events
- Display:
- Graphic illuminated and menu-guided navigation with up and down arrows
- Graphic illuminated and menu-guided navigation with up and down arrows
- Strøm:
- 6 V
- 6 V