Order Entry
Norway
ContactUsLinkComponent
VWR Collection Aluminum Weigh Dish, 34 mL, 57 x 14 mm, crinkle wall, with tab, pk100 1 * 100 items
Catalog # HEATHS14521BV
Supplier:  HEATHROW
New Product
VWR Collection Aluminum Weigh Dish, 34 mL, 57 x 14 mm, crinkle wall, with tab, pk100 1 * 100 items
Catalog # HEATHS14521BV
Supplier:  HEATHROW