0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Muffle Furnace LT15/14/C550 Tmax1400 with lift door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3442
Supplier:
NABERTHERM
New Product
Expand image
Muffle Furnace LT15/14/C550 Tmax1400 with lift door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3442
Supplier:
NABERTHERM
New Product