Order Entry
Norway
Orders LinkContactUsLinkComponent
Human Transferrin ELISA Kit

Human Transferrin ELISA Kit

Human Transferrin ELISA kit, 384 tests
Catalog # ABCAAB288174-1X384
New Product
Supplier:   Abcam
Human Transferrin ELISA Kit
Catalog # ABCAAB288174-1X384
Supplier:  Abcam
New Product
CAS Number:  
Human Transferrin ELISA Kit
Human Transferrin ELISA Kit

Human Transferrin ELISA kit, 384 tests

CAS Number:  
Price
kr 22 675,00
Log in for å se tilgjengelighet og deres pris
1 * 1 Each
Product Details & Documents

Human Transferrin ELISA kit is a single-wash 90-min Simplestep used to quantify Human Transferrin with a sensitivity of 4,396 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.

Spesifikasjoner

Spesifikasjoner

  • Description:
    Human Transferrin ELISA kit
  • Tests per kit:
    384 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Cell culture media, Heparin Plasma, Citrate plasma, Serum, EDTA Plasma
  • Detection range:
    15,625 - 1000 pg/ml
  • Storage temperature:
    4 ℃
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB288174-1X384
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    4,396 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Transferrin
  • Regulatory status:
    RUO

Spesifikasjoner

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.