0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Vials Snap Ring ND11, wide opening : 1.0ml Microliter con. Base , Snap Ring Vial, 32 x 11.6mm, amber glass, 1st hydrol. class 1 * 1.000 items
Catalog # LAPH11093406
Supplier:
LA PHA PACK
New Product
Expand image
Vials Snap Ring ND11, wide opening : 1.0ml Microliter con. Base , Snap Ring Vial, 32 x 11.6mm, amber glass, 1st hydrol. class 1 * 1.000 items
Catalog # LAPH11093406
Supplier:
LA PHA PACK
New Product