Order Entry
Norway
ContactUsLinkComponent
ECO 121-3 Device in carton box, test protocol, batteries, manual with immersion probe 100 mm, Ø 3 mm, Pt1000 2-wire fixed connection, V4A, 1m cable 1 * 1 items
Katalog # GRSG486750
Supplier:  Greisinger
New Product
ECO 121-3 Device in carton box, test protocol, batteries, manual with immersion probe 100 mm, Ø 3 mm, Pt1000 2-wire fixed connection, V4A, 1m cable 1 * 1 items
Katalog # GRSG486750
Supplier:  Greisinger
New Product