Order Entry
Norway
ContactUsLinkComponent
Scientific Line, Underbench ATEX Fridge Solid door, 3 Glass Shelves, temperature stability: +/-3°C, temperature range: +3/+16 °C, gr oss/net volume (L): 134/98, dimensions in mm (W/D/H): 597 / 605 / 819 1 * 1 items
Catalog # LIEBSRTFG1501
Supplier:  Liebherr
New Product
Scientific Line, Underbench ATEX Fridge Solid door, 3 Glass Shelves, temperature stability: +/-3°C, temperature range: +3/+16 °C, gr oss/net volume (L): 134/98, dimensions in mm (W/D/H): 597 / 605 / 819 1 * 1 items
Catalog # LIEBSRTFG1501
Supplier:  Liebherr