Order Entry
Norway
ContactUsLinkComponent
Rat SFXN1/TCC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat SFXN1/TCC in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270992-48
Supplier:  Antibodies.com
New Product
Rat SFXN1/TCC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat SFXN1/TCC in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270992-48
Supplier:  Antibodies.com