Order Entry
Norway
ContactUsLinkComponent
Human Factor X ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Factor X in plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1889-48
Supplier:  Antibodies.com
New Product
Human Factor X ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Factor X in plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1889-48
Supplier:  Antibodies.com