Order Entry
Norway
ContactUsLinkComponent
Mouse Heme Oxygenase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse Heme Oxygenase 1 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1787-48
Supplier:  Antibodies.com
New Product
Mouse Heme Oxygenase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse Heme Oxygenase 1 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1787-48
Supplier:  Antibodies.com