Order Entry
Norway
ContactUsLinkComponent
Rat IRS1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat IRS1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA270978-48
New Product
Rat IRS1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat IRS1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA270978-48