Mouse Factor X ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse Factor X in plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1891-48
Supplier: Antibodies.com