Order Entry
Norway
ContactUsLinkComponent
Rabbit MMP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit MMP2 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2426-48
Supplier:  Antibodies.com
New Product
Rabbit MMP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit MMP2 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2426-48
Supplier:  Antibodies.com