Order Entry
Norway
ContactUsLinkComponent
Human TXNIP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TXNIP in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2232-48
Supplier:  Antibodies.com
New Product
Human TXNIP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TXNIP in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2232-48
Supplier:  Antibodies.com