Order Entry
Norway
ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human GPCR GPR48/LGR4 protein to be purified and analysed in vitro. 1 * 10 µG
  ANTIA318435-10
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human GPCR GPR48/LGR4 protein to be purified and analysed in vitro. 1 * 10 µG
  ANTIA318435-10