Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human SLC1A5/ASCT2 protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA318446-100
Leverandør: Antibodies.com