Order Entry
Norway
ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human GPR2/CCR10 protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA325197-10
Leverandør:  Antibodies.com
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human GPR2/CCR10 protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA325197-10
Leverandør:  Antibodies.com