[EN]NANODISC HUMAN GPCR GPR124 PROTEIN 1 * 1 Each
Catalog # ANTIA325158-50
Supplier: Antibodies.com
New Product
[EN]NANODISC HUMAN GPCR GPR124 PROTEIN 1 * 1 Each
Catalog # ANTIA325158-50
Supplier: Antibodies.com
New Product
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Vann, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Hansker, nitril
Supplier: VWR Collection
VWR®, Pasteurpipetter, High-Performance
Supplier: VWR Collection
VWR®, Serologiske pipetter, Standard Line
Supplier: VWR Collection
Hansker, nitril, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Ansiktsservietter
Supplier: VWR Collection
Teststrips, pH, MQuant®, Supelco®
Supplier: Merck