Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Adenosine A1 Receptor protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA325085-10
Supplier: Antibodies.com