Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Frizzled 2/FZD2 protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325135-100
Supplier: Antibodies.com