Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human GPCR GPR37 protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325173-100
Supplier: Antibodies.com