Order Entry
Norway
ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human CRF1/CRHR1 protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA325112-10
Supplier:  Antibodies.com
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human CRF1/CRHR1 protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA325112-10
Supplier:  Antibodies.com