Order Entry
Norway
ContactUsLinkComponent
Human MT2-MMP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human MT2-MMP in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4460-48
Supplier:  ANTIBODIES.COM
New Product
Human MT2-MMP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human MT2-MMP in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4460-48
Supplier:  ANTIBODIES.COM
New Product