Order Entry
Norway
ContactUsLinkComponent
Human MT4-MMP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human MT4-MMP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Katalog # ANTIA8054-48
Supplier:  ANTIBODIES.COM
New Product
Human MT4-MMP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human MT4-MMP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Katalog # ANTIA8054-48
Supplier:  ANTIBODIES.COM
New Product