Order Entry
Norway
ContactUsLinkComponent
TuffStuff™ Nonwoven cellulose/polyester wipes, flat blue, 9 x 9 (23 x 23cm) 1 * 1 items
Catalog # CONTNWTS0001
Supplier:  CONTEC
New Product
TuffStuff™ Nonwoven cellulose/polyester wipes, flat blue, 9 x 9 (23 x 23cm) 1 * 1 items
Catalog # CONTNWTS0001
Supplier:  CONTEC
New Product