4-Hydroxyisoindolin-1-one
Bottle
Catalog # APOSOR305268-5G
New Product
Supplier: Apollo Scientific
4-Hydroxyisoindolin-1-one
Catalog # APOSOR305268-5G
Supplier: Apollo Scientific
New Product
CAS Number: 366453-21-6
Jump to:
Product Details & Documents
Synonyms: 2,3-Dihydro-4-hydroxy-1H-isoindol-1-one, 4-Hydroxy-2,3-dihydroisoindol-1-one, 2,3-Dihydro-4-hydroxy-1-oxo-1H-isoindole
- CAS Number:366453-21-6
- Formula:C₈H₇NO₂
- MDL Number:MFCD10000829
- Molecular Weight:149.147 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



