Order Entry
Norway
ContactUsLinkComponent
Horizontal Pre & Post Vacuum Autoclave. Pressure Vessel: chamber and door made of stainless steel 316L polished to 0.8 micron grade and electro- polished. The chamber is fully covered by jacket made of stainless steel 304L. Insulation cover and base 1 * 1 items
  TUTT5596-1V
 :  TUTTNAUER
Horizontal Pre & Post Vacuum Autoclave. Pressure Vessel: chamber and door made of stainless steel 316L polished to 0.8 micron grade and electro- polished. The chamber is fully covered by jacket made of stainless steel 304L. Insulation cover and base 1 * 1 items
  TUTT5596-1V
 :  TUTTNAUER