Buffer solution pH 9.00 ±0.01 (20 °C) 1 * 500 mL
ROTH1LP5.1
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ