Disposal Cabinet
Connecting frame for disposal cabinet/dispenser unit, 30×800×1600 mm
Catalog # PALBGDC8040-FRAME
Supplier: Palbam
Jump to:
Product Details & Documents
This disposal cabinet can be used as a stand-alone unit or in combination with the dispenser unit (connecting frame required).
- Made of stainless steel 304 with electropolished finish
- Two waste openings, two doors and levelling feet
- Does not require connecting to the wall
Can be used in cleanroom and critical environments.
: ISO certified.
: Typically available from stock for direct shipment from the supplier. Supplied with two large plastic bins.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifications
- Description:Connecting frame for disposal cabinet/dispenser unit
- Seq:01
- W×D×H:30×800×1600 mm