0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
5 components: Ca 2500mg/l Mg 500mg/l Na 1250mg/l S 1000mg/l Si 500mg/l in HNO3 2% 1 * 100 mL
Catalog # CMPLRM096157L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
5 components: Ca 2500mg/l Mg 500mg/l Na 1250mg/l S 1000mg/l Si 500mg/l in HNO3 2% 1 * 100 mL
Catalog # CMPLRM096157L1
Supplier:
CUSTOM MADE CHEMICALS LAB