Order Entry
Norway
ContactUsLinkComponent
Dimensions: 210 x 160 x 40 mm Color: PE natur Fluid: transparent white. Sealing:laminated aluminium film and protective cap (black).Will maintain a temperature between -23º C ... -19ºC . 1 * 1 items
Katalog # DLTTA100009
Supplier:  DELTA T
Dimensions: 210 x 160 x 40 mm Color: PE natur Fluid: transparent white. Sealing:laminated aluminium film and protective cap (black).Will maintain a temperature between -23º C ... -19ºC . 1 * 1 items
Katalog # DLTTA100009
Supplier:  DELTA T