kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Ferrous Ammonium Sulphate 0.1M Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 L
CMPLZ30054283
:
CUSTOM MADE CHEMICALS LAB
Ferrous Ammonium Sulphate 0.1M Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 L
CMPLZ30054283
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
: VWR Chemicals
pH indicator papers, universal/special
: MACHEREY-NAGEL
Hemacolor® Rapid staining of blood smear solution II staining reagent, Sigma-Aldrich®
: Merck
pH Papers, without color scale
: MACHEREY-NAGEL
Rose Bengal ≥80% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Ruthenium(III) chloride oxide, Technical Grade, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Polyvinyl alcohol, fully hydrolysed, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Test kits, chlorine (DPD), CHECKIT®
: Lovibond
Nitrite VHR L, 500 ml
: Lovibond