Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
40G PL SILICA LR 3 (190 TESTS) 1 * 40 g
Catalog # WTIDPLPOW40SILR3
Supplier:
WATER ID PRIMELAB
Expand image
40G PL SILICA LR 3 (190 TESTS) 1 * 40 g
Catalog # WTIDPLPOW40SILR3
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Acriflavine hydrochloride fluorescent dye, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Buffer concentrate, citrate phosphate, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Sodium hydroxide 98%, flakes, pure
Supplier: Thermo Fisher Scientific
6-CFDA (6-Carboxyfluorescein diacetate, single isomer), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY