Order Entry
Norway
ContactUsLinkComponent
Model 5075GS-2A Hospital grade pre and post vacuum autoclave, Diameter 50 cm Depth 81 cm. 160 Liter chamber. 316L grade stainless steel chamber with electro polish finish. Automatic locking door. Power: 400V 50/60Hz 3 Ph 18kW Built-in steam genera 1 * 1 items
  TUTT5075GS-2A-380
 :  TUTTNAUER
Model 5075GS-2A Hospital grade pre and post vacuum autoclave, Diameter 50 cm Depth 81 cm. 160 Liter chamber. 316L grade stainless steel chamber with electro polish finish. Automatic locking door. Power: 400V 50/60Hz 3 Ph 18kW Built-in steam genera 1 * 1 items
  TUTT5075GS-2A-380
 :  TUTTNAUER