kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
MYELIN BASIC PROTEIN (84-97) PEPTIDE 1MG, CRYOPRESERVED 1 * 1 mg
ANTIGEN-1053
:
Charles River Laboratories Cell Solutions
MYELIN BASIC PROTEIN (84-97) PEPTIDE 1MG, CRYOPRESERVED 1 * 1 mg
ANTIGEN-1053
:
Charles River Laboratories Cell Solutions