Order Entry
Norway
ContactUsLinkComponent
Smiths Medical® Fitting, Polycarbonate, Four-Way Stopcock, Male Slip Luer, Non-Sterile; 1000/Pk
Smiths Medical® Fitting, Polycarbonate, Four-Way Stopcock, Male Slip Luer, Non-Sterile; 1000/Pk
Catalog # MFLX30600-03BLK
undefined
Smiths Medical® Fitting, Polycarbonate, Four-Way Stopcock, Male Slip Luer, Non-Sterile; 1000/Pk
Catalog # MFLX30600-03BLK
About this item
Regulate flow without any leaking.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.
  • Flow pattern:
    Four-Way
  • Description:
    Fitting, Polycarbonate, Four-Way Stopcock, Male Slip Luer, Non-Sterile; 1000/Pk
  • Seq:
    05
  • Environmentally Preferable:
  • Wetted materials:
    Polycarbonate
  • Warranty:
    Out of box
  • Pk:
    1.000
  • Inlet port:
    Female Luer
  • Outlet port:
    Male Luer