9-Chloro-11beta,17,21-trihydroxy-16beta- methylpregna-1,4-diene-3 ,20-dione (Beclometasone) 1 * 25 mg
Catalog # PROOMM0199.03-0025
Supplier: LGC Standards PROMOCHEM
9-Chloro-11beta,17,21-trihydroxy-16beta- methylpregna-1,4-diene-3 ,20-dione (Beclometasone) 1 * 25 mg
Catalog # PROOMM0199.03-0025
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Propamocarb free base certified 1 * 0,1 g
Supplier: EHRENSTORFER
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
Pharmaceutical secondary standard, CMIT/MIT, (CRM), Supelco®
Supplier: Merck
Phosphatidic Acid Monosodium, Reference Standard
Supplier: USP
Alcohol calibration standards, Ethanol, (CRM), Supelco®
Supplier: Merck
VWR® ARISTAR®, Interference Check A in Nitric Acid 1% for ICP-MS
Supplier: VWR Chemicals
Organic reference standards, phenylephrine, Ph. Eur., RS
Supplier: EDQM
Total suspended solids calibration standards, Supelco®
Supplier: Merck
Trinexapac (free acid) 1 * 25 mg
Supplier: EHRENSTORFER
Diisobutyl phthalate
Supplier: EHRENSTORFER