Order Entry
Norway
ContactUsLinkComponent
Guardian 3000 Hotplate-Stirrer e-G31HSRDS 230V EU, Temperature Range: Ambient + 5°C – 380°C, Capacity 15L, Top Plate Dimensions: Round, 13.5 cm diameter, Ceramic Coated Stainless Steel 1 * 1 items
Catalog # OHAU30680278
Supplier:  OHAUS
Guardian 3000 Hotplate-Stirrer e-G31HSRDS 230V EU, Temperature Range: Ambient + 5°C – 380°C, Capacity 15L, Top Plate Dimensions: Round, 13.5 cm diameter, Ceramic Coated Stainless Steel 1 * 1 items
Catalog # OHAU30680278
Supplier:  OHAUS
Frequently Bought Together