Z-(L-ARG)-AMC (HYDROCHLORIDE) 1 * 50 mg
Catalog # CAYM30797-50
Supplier: Cayman Chemical
Frequently Bought Together
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Pyridine, EMPLURA®, Supelco®
Supplier: Merck
4-Methyl-2-pentanone ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Benzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
1,3-Dihydroxynaphthalene, Supelco®
Supplier: Merck
Chloroform 99+% stabilised for analysis
Supplier: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
2-Propanol, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific