Order Entry
Norway
ContactUsLinkComponent
CF and reg Dye Succinimidyl Esters (SE, also known as NHS esters) are Biotium's next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups, such as lysine residues. Red fluorescent CF and reg 5 1 * 1 µmol
Catalog # 96092.
Supplier:  Biotium
CF and reg Dye Succinimidyl Esters (SE, also known as NHS esters) are Biotium's next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups, such as lysine residues. Red fluorescent CF and reg 5 1 * 1 µmol
Catalog # 96092.
Supplier:  Biotium
Frequently Bought Together