Order Entry
Norway
ContactUsLinkComponent
RSA-Pro X Inserts, in Racks. Includes 96, Clear, 350ul Glass Low Volume Precision Inserts with wide Conical Points in plastic racks. U-2D Brand. 18 Racks / CS. 1 * 18 items
Catalog # MICS95296-350-P-CS
Supplier:  MICROSOLV
RSA-Pro X Inserts, in Racks. Includes 96, Clear, 350ul Glass Low Volume Precision Inserts with wide Conical Points in plastic racks. U-2D Brand. 18 Racks / CS. 1 * 18 items
Catalog # MICS95296-350-P-CS
Supplier:  MICROSOLV