Globe Stirrer Couplings, L 190 mm Ø 4 mm, with clamp screw for shaft O.D. 4 mm. Chuck adaptor O.D. 8 mm. 1 * 1 items
Catalog # BOHLC394-01
Supplier: Bohlender
Globe Stirrer Couplings, L 190 mm Ø 4 mm, with clamp screw for shaft O.D. 4 mm. Chuck adaptor O.D. 8 mm. 1 * 1 items
Catalog # BOHLC394-01
Supplier: Bohlender
Frequently Bought Together
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Magnetic hotplate stirrer, RH basic 2 IKAMAG®
Supplier: IKA
Vortex mixers, MS 3
Supplier: IKA
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
Overhead stirrer tools, 4-blade stirrers
Supplier: IKA
Accessories for magnetic hotplate stirrers
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for hotplates, VHP-C Series
Supplier: IKA
VWR®, Mini vortex mixer
Supplier: VWR Collection
VWR®, Magnetic Stirrer, Lab Disc
Supplier: VWR Collection
Vortex mixers, ZX4 and TX4
Supplier: VELP SCIENTIFIC
Programmable Vortex Mixers, Genie Series
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Accessories for Retsch sieves
Supplier: Retsch GmbH
Homogeniser, T 25 digital ULTRA-TURRAX®
Supplier: IKA
Electronic contact thermometers, ETS-D5/ ETS-D6
Supplier: IKA
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
Masterflex® Plastic Tubing Clamp Ties, Avantor®
Supplier: Avantor Fluid Handling