Order Entry
Norway
ContactUsLinkComponent
PERKINELMER’S HIGH THROUGHPUT SYSTEM (HTS) AFFORDS ULTRA-FAST ANALYSIS, DRAMATICALLY REDUCING LABORATORY OVERHEADS AND THE COST OF A NALYSIS. USING A METAL-FREE FLUID PATH, THE SYSTEM IS ABLE TO QUICKLY DELIVER THE SAMPLE TO PLASMA, WHILE HAVING UNCO 1 * 1 items
Catalog # PERCN0812026
Supplier:  PerkinElmer
PERKINELMER’S HIGH THROUGHPUT SYSTEM (HTS) AFFORDS ULTRA-FAST ANALYSIS, DRAMATICALLY REDUCING LABORATORY OVERHEADS AND THE COST OF A NALYSIS. USING A METAL-FREE FLUID PATH, THE SYSTEM IS ABLE TO QUICKLY DELIVER THE SAMPLE TO PLASMA, WHILE HAVING UNCO 1 * 1 items
Catalog # PERCN0812026
Supplier:  PerkinElmer
Frequently Bought Together