Order Entry
Norway
ContactUsLinkComponent
THE AUTOSAMPLER HAS A 3-CHANNEL PERISTALTIC PUMP THAT ACCEPTS BELOW LISTED TUBES WHEN RUNNING ORGANIC SOLVENTS. 2 - 2 MM ID VITON, B LACK TABS, RINSE 1 - 3.2MM ID VITON, WHITE TABS, WASTE 1 * 1 items
  PERCN0831039
THE AUTOSAMPLER HAS A 3-CHANNEL PERISTALTIC PUMP THAT ACCEPTS BELOW LISTED TUBES WHEN RUNNING ORGANIC SOLVENTS. 2 - 2 MM ID VITON, B LACK TABS, RINSE 1 - 3.2MM ID VITON, WHITE TABS, WASTE 1 * 1 items
  PERCN0831039