Order Entry
Norway
ContactUsLinkComponent
Ammonium Standard Solution with 50 mg/L NH?-N for quality control checks with LCK503 Ammonium cuvette test. LCA503 is also suitable for quality checks with the Amtax sc Ammonium analyser. 1 * 1 items
Catalog # HACHLCA503
Supplier:  Hach
Ammonium Standard Solution with 50 mg/L NH?-N for quality control checks with LCK503 Ammonium cuvette test. LCA503 is also suitable for quality checks with the Amtax sc Ammonium analyser. 1 * 1 items
Catalog # HACHLCA503
Supplier:  Hach
Frequently Bought Together