EPDM with Braiding Temperature range: -40°C…100°C Length: variable Max. pressure at 20°C: 20 bar Internal diameter: 12 mm 1 * 1 items
Katalog # HUBE10506
EPDM with Braiding Temperature range: -40°C…100°C Length: variable Max. pressure at 20°C: 20 bar Internal diameter: 12 mm 1 * 1 items
Katalog # HUBE10506