Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Megestrol Acetate, Reference Standards

Megestrol Acetate Related Compound C, USP reference standard, Vial, 20 mg
  1379128.
 :   USP
Megestrol Acetate, Reference Standards
  1379128.
 :  USP
 :  
Megestrol Acetate, Reference Standards

Megestrol Acetate Related Compound C, USP reference standard, Vial, 20 mg

 :  
 
kr 5 850,00
   
1 * 20 mg
 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

  • Pack type:
    Vial
  • Pk:
    20 mg
  • Standards:
    USP reference standard
  • Description:
    Megestrol Acetate Related Compound C
  • Cat. No.:
    1379128.
  • Description:
    Megestrol Acetate Related Compound C
  • Synonym(s):
    6,17a-β-Dimethyl-3,17-dioxo-17a-homoandrosta-4,6-diene-17a-α-yl acetate
  • CAS No.:
    74910-22-8